3-Amino-5-bromo-2-chloropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Bromo-2-chloro-3-pyridylamine
A2540-5G
A2540-25G
TCIAA2540-5GEA
91
EUR
TCIAA2540-5G
TCIAA2540-25G
3-Amino-5-bromo-2-chloropyridine ≥98.0% (by GC)
3-Amino-5-bromo-2-chloropyridine
Formula:
C₅H₄BrClN₂ Peso molecolare: 207,46 g/mol |
MDL Number:
MFCD02682092 CAS Number: 588729-99-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...