Stampa…

5-Amino-3-bromo-2-chloropyridine ≥95.0% (by GC)

Fornitore: TCI
Attenzione

A2539-1G A2539-5G
TCIAA2539-1GEA 26 EUR
TCIAA2539-1G TCIAA2539-5G
5-Amino-3-bromo-2-chloropyridine ≥95.0% (by GC)
5-Amino-3-bromo-2-chloropyridine
Formula: C₅H₄BrClN₂
Punto di fusione: 113°C
MDL Number: MFCD03840433
CAS Number: 130284-53-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR