Stampa…

3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 3-Amino-3-(3,4-dimethoxyphenyl)propionic acid

A2536-1G A2536-5G
TCIAA2536-1GEA 31 EUR
TCIAA2536-1G TCIAA2536-5G
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid
Formula: C₁₁H₁₅NO₄
Peso molecolare: 225,244 g/mol
MDL Number: MFCD00735172
CAS Number: 34841-09-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR