Amiodarone hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Amiodarone HCl, (2-Butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride)
A2530-1G
A2530-5G
TCIAA2530-1GEA
21
EUR
TCIAA2530-1G
TCIAA2530-5G
Amiodarone hydrochloride ≥98.0% (by HPLC, titration analysis)
Amiodarone hydrochloride
Formula:
C₂₅H₂₉I₂NO₃ Peso molecolare: 681,78 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00069204 CAS Number: 19774-82-4 EINECS: 243-293-2 Merck Index: 14,00482 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...