Stampa…

N-Allyl-N-tert-butylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-tert-Butylallylamine

A2514-25G A2514-100G
TCIAA2514-25GEA 182 EUR
TCIAA2514-25G TCIAA2514-100G
N-Allyl-N-tert-butylamine ≥98.0% (by GC, titration analysis)
N-Allyl-N-tert-butylamine
Formula: C₇H₁₅N
Peso molecolare: 113,20 g/mol
Punto di ebollizione: 112°C
CAS Number: 16486-68-3
UN: 2733
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR