Stampa…

L-Alanine ethyl ester hydrochloride ≥98.0% (by total nitrogen basis)

Fornitore: TCI

A2494-5G A2494-25G
TCIAA2494-5GEA 28 EUR
TCIAA2494-5G TCIAA2494-25G
L-Alanine ethyl ester hydrochloride ≥98.0% (by total nitrogen basis)
L-Alanine ethyl ester hydrochloride
Formula: C₅H₁₁NO₂
MDL Number: MFCD00063662
CAS Number: 1115-59-9
EINECS: 214-225-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 77.0 to 83.0 deg-C
Specific rotation [a]20/D +2.0 to +4.0 deg (C=2.5, H2O)

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR