Stampa…

1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI
Pericolo

A2493-1G A2493-5G
TCIAA2493-1GEA 28 EUR
TCIAA2493-1G TCIAA2493-5G
1-Amino-1-cyclopropanecarbonitrile hydrochloride ≥98.0% (by total nitrogen and titration analysis)
1-Amino-1-cyclopropanecarbonitrile hydrochloride
Formula: C₄H₇ClN₂
MDL Number: MFCD04114063
CAS Number: 127946-77-4
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR