Stampa…

Acarbose ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

A2485-1G A2485-5G
TCIAA2485-1GEA 59 EUR
TCIAA2485-1G TCIAA2485-5G
Acarbose ≥98.0% (by HPLC, total nitrogen)
Acarbose
Formula: C₂₅H₄₃NO₁₈
Peso molecolare: 645,61 g/mol
MDL Number: MFCD00869592
CAS Number: 56180-94-0
EINECS: 260-030-7
Merck Index: 14,00018

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +173 to +183 deg (C=1、H2O) (calcd.on anh.substance)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR