Stampa…

Aceclofenac ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(2-(2-((2,6-Dichlorophenyl)amino)phenyl)acetoxy)acetic acid, 2-[2-(2,6-Dichloroanilino)phenylacetoxy]acetic Acid

A2484-5G A2484-1G
TCIAA2484-5GEA 59 EUR
TCIAA2484-5G TCIAA2484-1G
Aceclofenac ≥98.0% (by HPLC, titration analysis)
Aceclofenac
Formula: C₁₆H₁₃Cl₂NO₄
Peso molecolare: 354,18 g/mol
MDL Number: MFCD00864296
CAS Number: 89796-99-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,00022

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR