Stampa…

Andrographolide 98,0 HPLC_ASSAY_METHOD

Fornitore: TCI

Synonyms: (S,E)-4-Hydroxy-3-(2-((1R,4aS,5R,6R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)dihydrofuran-2(3H)-one

A2459-1G A2459-5G
TCIAA2459-1GEA 44 EUR
TCIAA2459-1G TCIAA2459-5G
Andrographolide 98,0 HPLC_ASSAY_METHOD
Andrographolide
Formula: C₂₀H₃₀O₅
Peso molecolare: 350.45 g/mol
MDL Number: MFCD07778082
CAS Number: 5508-58-7
EINECS: 226-852-5
Merck Index: 14,00633

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 229.0 to 235.0 deg-C
Specific rotation [a]20/D -124 to -129 deg (C=1.5, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR