1-Amino-2-methylpropan-2-ol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₄H₁₁NO Punto di ebollizione: 151°C |
MDL Number:
MFCD01697283 CAS Number: 2854-16-2 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...