Stampa…

4-Amino-6-hydroxypyrimidine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A2439-1G A2439-5G
TCIAA2439-1GEA 69 EUR
TCIAA2439-1G TCIAA2439-5G
4-Amino-6-hydroxypyrimidine ≥98.0% (by titrimetric analysis)
4-Amino-6-hydroxypyrimidine
Formula: C₄H₅N₃O
Peso molecolare: 111,10 g/mol
Punto di fusione: 265°C
MDL Number: MFCD00051502
CAS Number: 1193-22-2
EINECS: 214-771-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR