Stampa…

5-Amino-1-methyltetrazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-1H-tetrazol-5-amine

A2427-25G A2427-5G
TCIAA2427-25GEA 193 EUR
TCIAA2427-25G TCIAA2427-5G
5-Amino-1-methyltetrazole ≥98.0% (by GC, titration analysis)
5-Amino-1-methyltetrazole
Formula: C₂H₅N₅
Peso molecolare: 99,10 g/mol
MDL Number: MFCD00186454
CAS Number: 5422-44-6
UN: 1325
ADR: 4.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR