Stampa…

Amezinium methyl sulphate ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

Synonyms: 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate, Amezinium Methyl sulphate, Lu-1631, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulphate, Amezinium methylsulphate, Amezinium metilsulfate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulphate

A2425-1G A2425-5G
TCIAA2425-1GEA 53 EUR
TCIAA2425-1G TCIAA2425-5G
Amezinium methyl sulphate ≥98.0% (by HPLC)
Amezinium methyl sulphate
Formula: C₁₂H₁₅N₃O₅S
Peso molecolare: 313.33 g/mol
Punto di fusione: 176°C
MDL Number: MFCD00865972
CAS Number: 30578-37-1
EINECS: 250-248-0
Merck Index: 14,00394

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.10 to 13.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR