Stampa…

N-(4-Aminobutyl)-2-naphthalenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: W-12 Hydrochloride

A2408-25MG A2408-500MG
TCIAA2408-25MGEA 16 EUR
TCIAA2408-25MG TCIAA2408-500MG
N-(4-Aminobutyl)-2-naphthalenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
N-(4-Aminobutyl)-2-naphthalenesulphonamide hydrochloride
Formula: C₁₄H₁₈N₂O₂S
Peso molecolare: 314,83 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00058052
CAS Number: 89108-46-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 181.0 to 185.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR