Atropic acid ≥93.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2-Phenylacrylic Acid, Ipratropium bromide Impurity D
Formula:
C₉H₈O₂ Punto di ebollizione: 267°C |
MDL Number:
MFCD00046531 CAS Number: 492-38-6 EINECS: 207-753-6 Merck Index: 14,00874 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...