Stampa…

Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI

A2360-25G A2360-5G
TCIAA2360-25GEA 281 EUR
TCIAA2360-25G TCIAA2360-5G
Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ambroxol hydrochloride
Formula: C₁₃H₁₈Br₂N₂O.HCl
Punto di fusione: 235°C
MDL Number: MFCD00078932
CAS Number: 23828-92-4
EINECS: 245-899-2
Merck Index: 14,00386

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR