Stampa…

2-Amino-3-pyridinemethanol ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-3-hydroxymethylpyridine, 2-Amino-3-(hydroxymethyl)pyridine

A2358-1G A2358-5G
TCIAA2358-1GEA 69 EUR
TCIAA2358-1G TCIAA2358-5G
2-Amino-3-pyridinemethanol ≥96.0% (by GC)
2-Amino-3-pyridinemethanol
Formula: C₆H₈N₂O
Peso molecolare: 124,14 g/mol
MDL Number: MFCD05663510
CAS Number: 23612-57-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 65.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR