Stampa…

Atomoxetine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (R)-N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine Hydrochloride, (R)-Tomoxetine Hydrochloride, (R)-N-Methyl-3-phenyl-3-(o-tolyloxy)propan-1-amine hydrochloride

A2357-1G A2357-100MG
TCIAA2357-1GEA 94 EUR
TCIAA2357-1G TCIAA2357-100MG
Atomoxetine hydrochloride ≥98.0% (by HPLC, titration analysis)
Atomoxetine hydrochloride
Formula: C₁₇H₂₁NO
Peso molecolare: 291,82 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD06410992
CAS Number: 82248-59-7
Merck Index: 14,00863

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C
Specific rotation [a]20/D -41.0 to -44.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR