Stampa…

Amlodipine besylate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

Synonyms: 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylic acid 3-Ethyl 5-Methyl Ester Benzenesulphon, 3-Ethyl 5-Methyl 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Benzenesulphonate, 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate benzenesulphonate

A2353-1G A2353-5G
TCIAA2353-1GEA 49 EUR
TCIAA2353-1G TCIAA2353-5G
Amlodipine besylate ≥98.0% (by HPLC, total nitrogen)
Amlodipine besylate
Formula: C₂₀H₂₅ClN₂O₅
Peso molecolare: 567.05 g/mol
Punto di fusione: 200°C
MDL Number: MFCD00887594
CAS Number: 111470-99-6
Merck Index: 14,00491

Visualizza i codici

Risultati del test di specificazione

Appearance White to Dark green powder to crystal
Purity (HPLC) min. 98.0 % (total of isomer)
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR