Stampa…

4-Amino-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: 4-Amino-o-toluic acid, 4-Amino-2-methylbenzoic Acid

A2349-25G A2349-5G
TCIAA2349-25GEA 89 EUR
TCIAA2349-25G TCIAA2349-5G
4-Amino-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-2-methylbenzoic acid
Formula: C₈H₉NO₂
MDL Number: MFCD06656133
CAS Number: 2486-75-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR