Stampa…

(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride, (S)-1-Cbz-3-Aminopyrrolidine hydrochloride

A2324-1G A2324-5G
TCIAA2324-1GEA 107 EUR
TCIAA2324-1G TCIAA2324-5G
(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride
Formula: C₁₂H₁₆N₂O₂
Peso molecolare: 256,73 g/mol
MDL Number: MFCD06796559
CAS Number: 550378-39-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +13.0 to +16.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR