Stampa…

(1R,2R)-(-)-1-Amino-2-indanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (1R,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol, (1R,2R)-(-)-1-Amino-2-hydroxyindan, (1R,2R)-1-Aminoindan-2-ol

A2306-1G A2306-5G
TCIAA2306-1GEA 63 EUR
TCIAA2306-1G TCIAA2306-5G
(1R,2R)-(-)-1-Amino-2-indanol ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1-Amino-2-indanol
Formula: C₉H₁₁NO
Peso molecolare: 149,19 g/mol
MDL Number: MFCD08275447
CAS Number: 163061-73-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR