Stampa…

3-Amino-2-pyridone ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone

A2285-1G A2285-5G
TCIAA2285-1GEA 75 EUR
TCIAA2285-1G TCIAA2285-5G
3-Amino-2-pyridone ≥96.0% (by GC, titration analysis)
3-Amino-2-pyridone
Formula: C₅H₆N₂O
Peso molecolare: 110,12 g/mol
MDL Number: MFCD03411556
CAS Number: 33630-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00acc410b715891a93138653edf8e0ca44c0ff01&vid=5cb96a0c7d9c6aa28bd84fb3ce5e289d42ee449a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystalline
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR