5-Chloro-2-(4-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-Amino-4,4'-dichlorodiphenyl ether, 5-Chloro-2-(4-chlorophenoxy)aniline
A2225-25G
A2225-5G
TCIAA2225-25GEA
195
EUR
TCIAA2225-25G
TCIAA2225-5G
5-Chloro-2-(4-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
5-Chloro-2-(4-chlorophenoxy)aniline
Formula:
C₁₂H₉Cl₂NO Peso molecolare: 254,1100 g/mol Punto di ebollizione: 234°C |
MDL Number:
MFCD00027391 CAS Number: 121-27-7 EINECS: 204-460-5 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...