Ammonium thioglycolate ∼60% in water
Fornitore: TCI
|
Pericolo
|
Synonyms:
Ammonium thioglycolate solution, Thioglycolic Acid Ammonium Salt, Ammonium Mercaptoacetate
Formula:
C₂H₇NO₂S Peso molecolare: 109.14 g/mol |
MDL Number:
MFCD00137451 CAS Number: 5421-46-5 EINECS: 226-540-9 UN: 2922 ADR: 8 III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Iodometric titration) | 57.0 to 63.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...