Stampa…

3-Amino-N-methylbenzylamine ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

A2158-25G A2158-5G
TCIAA2158-25GEA 41 EUR
TCIAA2158-25G TCIAA2158-5G
3-Amino-N-methylbenzylamine ≥97.0% (by GC, titration analysis)
3-Amino-N-methylbenzylamine
Formula: C₈H₁₂N₂
MDL Number: MFCD00211072
CAS Number: 18759-96-1
EINECS: 414-570-7
UN: 3259
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Green to Brown powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR