4-((1H-1,2,4-Triazol-1-yl)methyl)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A2148-1G
A2148-5G
TCIAA2148-1GEA
16
EUR
TCIAA2148-1G
TCIAA2148-5G
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline ≥98.0% (by GC, titration analysis)
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline
Formula:
C₉H₁₀N₄ Peso molecolare: 174,21 g/mol |
MDL Number:
MFCD00973306 CAS Number: 119192-10-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...