Stampa…

N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Amino-4,6-dichloro-5-formamidopyrimidine, N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide

A2146-25G A2146-5G
TCIAA2146-25GEA 127 EUR
TCIAA2146-25G TCIAA2146-5G
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98.0% (by HPLC, total nitrogen)
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide
Formula: C₅H₄Cl₂N₄O
Peso molecolare: 207,0200 g/mol
MDL Number: MFCD04112936
CAS Number: 171887-03-9
EINECS: 425-650-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR