Stampa…

3-Acetamidotetrahydro-2-thiophenone ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: Citiolone, 2-Acetamido-4-mercaptobutyric Acid gamma-Thiolactone, N-Acetyl-DL-homocystein Thiolactone, N-(Tetrahydro-2-oxo-3-thienyl)acetamide, N-(2-Oxotetrahydrothiophen-3-yl)acetamide

A2144-25G A2144-5G
TCIAA2144-25GEA 176 EUR
TCIAA2144-25G TCIAA2144-5G
3-Acetamidotetrahydro-2-thiophenone ≥98.0% (by GC, total nitrogen)
3-Acetamidotetrahydro-2-thiophenone
Formula: C₆H₉NO₂S
Peso molecolare: 159,20 g/mol
MDL Number: MFCD00005480
CAS Number: 1195-16-0
EINECS: 214-793-8
Merck Index: 14,02320

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR