Stampa…

2-Adamantanone oxime ≥98.0% (by GC)

Fornitore: TCI

A2110-25G A2110-5G
TCIAA2110-25GEA 164 EUR
TCIAA2110-25G TCIAA2110-5G
2-Adamantanone oxime ≥98.0% (by GC)
2-Adamantanone oxime
Formula: C₁₀H₁₅NO
MDL Number: MFCD00078273
CAS Number: 4500-12-3
EINECS: 224-807-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR