Azathioprine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
6-((1-Methyl-4-nitro-1H-imidazol-5-yl)thio)-9H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine
A2069-25G
A2069-5G
TCIAA2069-25GEA
368
EUR
TCIAA2069-25G
TCIAA2069-5G
Azathioprine ≥98.0% (by HPLC, titration analysis)
Azathioprine
Formula:
C₉H₇N₇O₂S Peso molecolare: 277,26 g/mol Punto di fusione: 244°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00069203 CAS Number: 446-86-6 EINECS: 207-175-4 Merck Index: 14,00902 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...