Stampa…

D-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Arg-OMe.2HCl

A2016-1G A2016-5G
TCIAA2016-1GEA 43 EUR
TCIAA2016-1G TCIAA2016-5G
D-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
D-Arginine methyl ester dihydrochloride
Formula: C₇H₁₆N₄O₂
Peso molecolare: 261,15 g/mol
MDL Number: MFCD00153417
CAS Number: 78851-84-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -22.0 deg (C=2.5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR