Ala-OBzl TosOH ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₁₇H₂₁NO₅S |
MDL Number:
MFCD00066143 CAS Number: 42854-62-6 EINECS: 255-969-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Specific rotation [a]20/D | -4.0 to -7.0 deg (C=3, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...