Stampa…

D-Alanine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Ala-OBzl.TosOH, H-D-Ala-OBzl·TosOH, H-D-Ala-OBzl.Tos-OH

A2004-25G A2004-5G
TCIAA2004-25GEA 244 EUR
TCIAA2004-25G TCIAA2004-5G
D-Alanine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
D-Alanine benzyl ester-p-toluenesulphonate
Formula: C₁₀H₁₃NO₂
Peso molecolare: 351,42 g/mol
MDL Number: MFCD00039092
CAS Number: 41036-32-2
EINECS: 255-185-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Specific rotation [a]20/D +4.0 to +7.0 deg (C=3, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR