6-Amino-2,4-dichloro-3-ethylphenol hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A2001-25G
A2001-5G
TCIAA2001-25GEA
138
EUR
TCIAA2001-25G
TCIAA2001-5G
6-Amino-2,4-dichloro-3-ethylphenol hydrochloride ≥98.0% (by HPLC, titration analysis)
6-Amino-2,4-dichloro-3-ethylphenol hydrochloride
Formula:
C₈H₉Cl₂NO Peso molecolare: 242,52 g/mol |
CAS Number:
101819-99-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...