N-(4-Oxocyclohexyl)acetamide ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Acetamidocyclohexanone
A1929-25G
A1929-5G
TCIAA1929-25GEA
47
EUR
TCIAA1929-25G
TCIAA1929-5G
N-(4-Oxocyclohexyl)acetamide ≥98.0% (by GC)
N-(4-Oxocyclohexyl)acetamide
Formula:
C₈H₁₃NO₂ Peso molecolare: 155,20 g/mol |
MDL Number:
MFCD03703462 CAS Number: 27514-08-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...