Stampa…

(S)-2-Aminopropanamide hydrochloride ≥97.0% (by total nitrogen basis)

Fornitore: TCI

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

A1919-25G A1919-5G
TCIAA1919-25GEA 72 EUR
TCIAA1919-25G TCIAA1919-5G
(S)-2-Aminopropanamide hydrochloride ≥97.0% (by total nitrogen basis)
(S)-2-Aminopropanamide hydrochloride
Formula: C₃H₉ClN₂O
MDL Number: MFCD00066145
CAS Number: 33208-99-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Specific rotation [a]20/D +9.0 to +13.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR