Stampa…

6-Acrylamidohexanoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

A1896-1G A1896-25G A1896-5G
TCIAA1896-1GEA 59 EUR
TCIAA1896-1G TCIAA1896-25G TCIAA1896-5G
6-Acrylamidohexanoic acid ≥98.0% (by titrimetric analysis)
6-Acrylamidohexanoic acid
Formula: C₉H₁₅NO₃
MDL Number: MFCD06804452
CAS Number: 20766-85-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR