2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Acetyl-5,5-dimethylcyclohexane-1,3-dione, 2-Acetyldimedone
A1853-1G
A1853-5G
TCIAA1853-1GEA
44
EUR
TCIAA1853-1G
TCIAA1853-5G
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98.0% (by GC)
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione
Formula:
C₁₀H₁₄O₃ Peso molecolare: 182,22 g/mol |
MDL Number:
MFCD00467663 CAS Number: 1755-15-3 |
Risultati del test di specificazione
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...