2-Amino-6-chloro-3-nitropyridine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₄ClN₃O₂ |
MDL Number:
MFCD00044350 CAS Number: 27048-04-0 EINECS: 248-188-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...