Stampa…

9-Aminoanthracene ≥96.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-Anthramine

A1748-100MG A1748-1G A1748-5G
TCIAA1748-100MGEA 51 EUR
TCIAA1748-100MG TCIAA1748-1G TCIAA1748-5G
9-Aminoanthracene ≥96.0% (by GC, total nitrogen)
9-Aminoanthracene
Formula: C₁₄H₁₁N
Peso molecolare: 193,25 g/mol
Punto di fusione: 170°C
MDL Number: MFCD00452690
CAS Number: 779-03-3

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Brown to Dark green powder to crystal
Purity (GC) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR