Adamantan-1-yl acrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Acrylic acid Adamantan-1-yl ester, 1-Acryloyloxyadamantane
Stabilization:
stabilizzato con BHT/Ionolo
A1735-1G
A1735-5G
TCIAA1735-1GEA
90
EUR
TCIAA1735-1G
TCIAA1735-5G
Adamantan-1-yl acrylate ≥98.0% (by GC) stabilizzato
Adamantan-1-yl acrylate
Formula:
C₁₃H₁₈O₂ Peso molecolare: 206,29 g/mol |
MDL Number:
MFCD18072700 CAS Number: 121601-93-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...