1-Adamantyl isocyanate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Isocyanic acid 1-Adamantyl Ester, 1-Adamantyl Isocyanate
Formula:
C₁₁H₁₅NO |
MDL Number:
MFCD00134405 CAS Number: 4411-25-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...