Stampa…

4-Amino-4'-dimethylaminoazobenzene ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

A1672-1G A1672-5G
TCIAA1672-1GEA 104 EUR
TCIAA1672-1G TCIAA1672-5G
4-Amino-4'-dimethylaminoazobenzene ≥97.0% (by HPLC, titration analysis)
4-Amino-4'-dimethylaminoazobenzene
Formula: C₁₄H₁₆N₄
MDL Number: MFCD00010204
CAS Number: 539-17-3
EINECS: 208-712-5

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to Amber to Dark red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 187.0 to 191.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR