4-(4-Aminopiperidino)pyridine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
4-Amino-1-(4-pyridyl)piperidine Dihydrochloride, 4-(4-Aminopiperidino)pyridine Dihydrochloride
Formula:
C₁₀H₁₅N₃ Peso molecolare: 250.17 g/mol |
MDL Number:
MFCD06797043 CAS Number: 1169396-92-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...