(S)-2-Aminopent-4-enoic acid hydrochloride ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
H-Allyl-Gly-OH·HCl, L-2-Allylglycine hydrochloride, H-Gly(Ally)-OH HCl, L-Allylglycine hydrocholride
A1648-1G
A1648-200MG
A1648-5G
TCIAA1648-1GEA
121
EUR
TCIAA1648-1G
TCIAA1648-200MG
TCIAA1648-5G
(S)-2-Aminopent-4-enoic acid hydrochloride ≥97.0% (by titrimetric analysis)
(S)-2-Aminopent-4-enoic acid hydrochloride
Formula:
C₅H₁₀ClNO₂ Peso molecolare: 151,59 g/mol |
MDL Number:
MFCD06797042 CAS Number: 195316-72-4 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Specific rotation [a]20/D | -8.0 to -13.0 deg (C=2, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...