1-Acetyl-1,2,3,4-tetrahydroquinoline ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₃NO Punto di ebollizione: 295°C |
MDL Number:
MFCD01195305 CAS Number: 4169-19-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...