Dimethyl azobenzene-4,4'-dicarboxylate ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Dimethyl 4,4'-Azodibenzoate, 4,4'-Azodibenzoic Acid Dimethyl Ester, Azobenzene-4,4'-dicarboxylic Acid Dimethyl Ester
A1581-1G
A1581-5G
TCIAA1581-1GEA
105
EUR
TCIAA1581-1G
TCIAA1581-5G
Dimethyl azobenzene-4,4'-dicarboxylate ≥95.0% (by GC)
Dimethyl azobenzene-4,4'-dicarboxylate
Formula:
C₁₆H₁₄N₂O₄ Peso molecolare: 298,30 g/mol |
MDL Number:
MFCD00496622 CAS Number: 5320-91-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 238.0 to 243.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...