Stampa…

2-Amino-4-methoxy-6-methyl-1,3,5-triazine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A1560-25G A1560-5G
TCIAA1560-25GEA 51 EUR
TCIAA1560-25G TCIAA1560-5G
2-Amino-4-methoxy-6-methyl-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2-Amino-4-methoxy-6-methyl-1,3,5-triazine
Formula: C₅H₈N₄O
Peso molecolare: 140,1400 g/mol
MDL Number: MFCD00052764
CAS Number: 1668-54-8
EINECS: 216-790-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 1mol/L HCl almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR